missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NOX3 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
- A blocking peptide containing 15 amino acids near the amino terminus of human NOX3
- NOX3 peptide is used for blocking the NOX3 antibody, NBP1-41292.
- Source: Synthetic
- Type: Blocking Peptide
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| Gene ID (Entrez) | 50508 |
| Protein Family | NOX3 |
| Concentration | 0.2 mg/mL |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | NOX3 |
| Quantity | 0.05 mg |
| Source | Synthetic |
| Storage Buffer | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction