missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nox4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-58851MFV610
This item is not returnable.
View return policy
Description
Nox4 Polyclonal antibody specifically detects Nox4 in Human, Mouse, Rat, Porcine, Primate samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| Nox4 | |
| Polyclonal | |
| Flow Cytometry, Flow (Intracellular) | |
| EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- | |
| A synthetic peptide made to a C-terminal region (within residues 500-578) of the human NOX4 protein sequence. [Swiss-Prot# Q9NPH5]. | |
| 0.1 mL | |
| Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
| 50507 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction