Learn More
Invitrogen™ NOX5 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA595572
Product Code. 16384835
  • € 530.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human placenta tissue, human SGC-7901 whole cell, human Jurkat whole cell, human THP-1 whole cell. ICC/IF: Hela cell. Flow: PC-3. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene is predominantly expressed in the testis and lymphocyte-rich areas of spleen and lymph nodes. It encodes a calcium-dependen NADPH oxidase that generates superoxide, and functions as a calcium-dependent proton channel that may regulate redox-dependent processes in lymphocytes and spermatozoa. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Oct 2011].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NOX5
Polyclonal
Unconjugated
NOX5
NADPH oxidase 5; NADPH oxidase, EF-hand calcium binding domain 5; NOX5; NOX5zeta
Rabbit
Affinity chromatography
RUO
79400
-20°C
Lyophilized
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
500 μg/mL
PBS with 4mg trehalose and 0.05mg sodium azide
Q96PH1
NOX5
E.coli-derived human NOX5 recombinant protein (Position: E13-F197).