missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noxa Antibody (114C307.1R) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07012SS
This item is not returnable.
View return policy
Description
Noxa Monoclonal antibody specifically detects Noxa in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular)
Specifications
| Noxa | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 μg/ml, Flow Cytometry, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1-5 μg/ml, Flow (Intracellular) | |
| adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
| This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa. | |
| 0.025 mg | |
| Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Mitochondrial Mediated Pathway, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
| 5366 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 114C307.1R | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction