missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOXRED1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99260-100ul
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
NOXRED1 Polyclonal antibody specifically detects NOXRED1 in Human samples. It is validated for Western Blot
Tekniske data
| NOXRED1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| C14orf148, NADP-dependent oxidoreductase domain containing 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NOXRED1. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 122945 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion