missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NP, Clone: B1-8, Abnova™
Monoclonal Antibody
Brand: Abnova RAB00157.200ug
This item is not returnable.
View return policy
Specifications
| NP | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.02% Proclin 300). | |
| 200 μg | |
| Primary | |
| Antibody | |
| IgG1 λ |
| Flow Cytometry, Western Blot | |
| B1-8 | |
| Flow Cytometry (1 ug/mL) Negative Control Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Original antibody is raised against hapten-protein conjugated NP12-chicken gammaglobulin. | |
| RUO | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction