missing translation for 'onlineSavingsMsg'
Learn More

NP118809, MedChemExpress

Product Code. p-200095880
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16446476

Brand: Medchem Express HY14462

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NP118809 is a potent N-type calcium channel blocker, with an IC50 of 0.11 μM; also less potently inhibits L-type calcium channel with an IC50 of 12.2 μM.

NP118809 is a potent N-type calcium channel blocker, with an IC50 of 0.11 μM

Specifications

Chemical Name or Material NP118809
CAS 41332-24-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C32H32N2O
Quantity 100 mg
Synonym 39-1B4
Solubility Information DMSO : 50 mg/mL (108.55 mM; Need ultrasonic)
SMILES O=C(CC(C1=CC=CC=C1)C2=CC=CC=C2)N3CCN(C(C4=CC=CC=C4)C5=CC=CC=C5)CC3
Molecular Weight (g/mol) 460.61
Formula Weight 460.61
Percent Purity 98.79%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.