missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPAS1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16373585
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16373585 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16373585 Supplier Invitrogen™ Supplier No. PA586721

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPAS1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NPAS1
Gene Accession No. P97459, Q99742
Gene Alias basic-helix-loop-helix-PAS protein MOP5; BHLHE11; Class E basic helix-loop-helix protein 11; member of PAS protein 5; member of PAS superfamily 5; MOP5; neuronal PAS domain protein 1; neuronal PAS domain-containing protein 1; Neuronal PAS1; Npas1; PAS domain-containing protein 5; PASD5
Gene Symbols NPAS1
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 486-531 of Human NPAS1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18142, 308387, 4861
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.