missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Npas4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP2-47252
This item is not returnable.
View return policy
Description
Npas4 Polyclonal specifically detects Npas4 in Mouse, Rat, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Npas4 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris 0.1M Glycine and 2% Sucrose with No Preservative | |
| NPAS4 | |
| Synthetic peptide derived from Cter domain of mouse Npas4 protein. | |
| 0.1 mg | |
| Primary | |
| Reconstitute with distilled water. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:1000 - 1:5000, ELISA, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:100 - 1:1000 | |
| BHLHE79, bHLHe79neuronal PAS domain-containing protein 4, Class E basic helix-loop-helix protein 79, Le-PAS, neuronal PAS domain protein 4, Neuronal PAS4, NXFHLH-PAS transcription factor NXF, PASD10PAS domain-containing protein 10 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 266743 | |
| Mouse, Rat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction