Learn More
Invitrogen™ NPAS4 Monoclonal Antibody (N408/79), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545750
Description
A 1:100 dilution of MA5-45750 was sufficient for detection of NPAS4 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 90kDa.approximately This antibody was formerly sold as clone S408-79.
NXF is a member of the basic helix-loop-helix-PER (MIM 602260)-ARNT (MIM 126110)-SIM (MIM 600892) (bHLH-PAS) class of transcriptional regulators, which are involved in a wide range of physiologic and developmental events.
Specifications
| NPAS4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| Q8BGD7, Q8CJH6, Q8IUM7 | |
| NPAS4 | |
| Fusion protein amino acids 597-802 (C-terminus) of rat Npas4. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N408/79 | |
| FITC | |
| NPAS4 | |
| bHLH PAS type transcription factor NXF; bHLHe79; bHLH-PAS type transcription factor NXF; Class E basic helix-loop-helix protein 79; HLH-PAS transcription factor NXF; LE-PAS; Limbic-enhanced PAS protein; neuronal PAS domain protein 4; neuronal PAS domain-containing protein 4; Neuronal PAS4; Npas4; NXF; PAS domain-containing protein 10; PASD10; PASD10 and LE-PAS | |
| Mouse | |
| Protein G | |
| RUO | |
| 225872, 266734, 266743 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.