missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPDC1 Monoclonal Antibody (16E4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA112777
This item is not returnable.
View return policy
Description
A suggested positive control for this product is brain tissue lysate.
The discovery of NPDC-1 (neuronal proliferation, differentiation and control protein 1) was the result of a study of genes that were preferentially expressed in immortalized neural precursor cell lines at the onset of contact inhibition of proliferation and subsequent differentiation. It was further found that transfection of transformed cells with NPDC-1 led to increased doubling times and suppression of many characteristics of transformation. NPDC-1 is differentially expressed in brain and lung and has been shown to interact directly with the transcription factor E2F-1 as well as D-type cyclins and cdk2. NPDC-1 has been identified as a modulator of transcriptiona levents mediated by retinoic acid and the protein level and activity of NPDC-1 have been shown to be regulated by phosphorylation mediated proteasomal degradation.
Specifications
| NPDC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 3mM sodium azide; pH 7.5 | |
| Q64322, Q9NQX5 | |
| NPDC1 | |
| Synthetic peptide corresponding to human NPDC-1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 16E4 | |
| Unconjugated | |
| NPDC1 | |
| AI314472; CAB; CAB-; CAB1; CAB-1; DKFZP586J0523; Neural proliferation differentiation and control protein 1; neural proliferation differentiation control-1 protein; neural proliferation, differentiation and control 1; neural proliferation, differentiation and control gene 1; neural proliferation, differentiation and control, 1; NPDC1; NPDC-1; RP23-47P18.6 | |
| Mouse | |
| Ion-exchange Chromatography | |
| RUO | |
| 18146, 296562, 56654 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction