Learn More
Invitrogen™ NPEPL1 Monoclonal Antibody (3F8-1A6)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA522592
Product Code. 15559142
  • € 534.00 / 100µg
Description
Peptide Sequence: MANVGLQFQA SAGDSDPQSR PLLLLGQLHH LHRVPWSHVR GKLQPRVTEE LWQAALSTLN PNPTDSCPLY LNYATVAALP CRVSRHNSPS AAHFITRLVR TCLPPGAHRC IVMVCEQPEV FASACALARA FPLFTHRSGA SRRLEKKTVT VEFFLVGQDN GPVEVSTLQC LANATDGVRL AARIVDTPCN EMNTDTFLEE INKVGKELGI IPTIIRDEEL KTRGFGGIYG VGKAALHPPA LAVLSHTPDG ATQTIAWVGK GIVYDTGGLS IKGKTTMPGM KRDCGGAAAV LGAFRAAIKQ GFKDNLHAVF CLAENSVGPN ATRPDDIHLL YSGKTVEINN TDAEGRLVLA DGVSYACKDL GADIILDMAT LTGAQGIATG KYHAAVLTNS AEWEAACVKA GRKCGDLVHP LVYCPELHFS EFTSAVADMK NSVADRDNSP SSCAGLFIAS HIGFDWPGVW VHLDIAAPVH AGERATGFGV ALLLALFGRA SEDPLLNLVS PLGCEVDVEE GDLGRDSKRR RLV.
Probably catalyzes the removal of unsubstituted N-terminal amino acids from various peptides.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NPEPL1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8NDH3
NPEPL1
NPEPL1 (AAH20507, 1 a.a. ~ 523 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG3 κ
ELISA, Western Blot, Immunocytochemistry
3F8-1A6
Unconjugated
NPEPL1
Aminopeptidase-like 1; bA261P9.2; BC023239; C85514; KIAA1974; mKIAA1974; NPEPL1; Probable aminopeptidase NPEPL1
Mouse
Affinity chromatography
RUO
79716
-20°C, Avoid Freeze/Thaw Cycles