missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPEPPS Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121735
This item is not returnable.
View return policy
Description
NPEPPS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes the puromycin-sensitive aminopeptidase, a zinc metallopeptidase which hydrolyzes amino acids from the N-terminus of its substrate. The protein has been localized to both the cytoplasm and to cellular membranes. This enzyme degrades enkaphalins in the brain, and studies in mouse suggest that it is involved in proteolytic events regulating the cell cycle.
Specifications
| NPEPPS | |
| Polyclonal | |
| Unconjugated | |
| NPEPPS | |
| AAP-S; aminopeptidase puromycin sensitive; cytosol alanyl aminopeptidase; goku; metalloproteinase MP100; MP100; NPEPPS; Psa; Puromycin-sensitive aminopeptidase; R74825 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 414413, 9520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P55786 | |
| NPEPPS | |
| Recombinant protein Puromycin Sensitive Aminopeptidase. The antigen corresponds to amino acid range Gly584-Gln793 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction