missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPFF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 463.00
Specifications
| Antigen | NPFF |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653139
|
Novus Biologicals
NBP3-05565-100ul |
100 μg |
€ 463.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660068
|
Novus Biologicals
NBP3-05565-20ul |
20 μg |
€ 188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NPFF Polyclonal antibody specifically detects NPFF in Human, Rat samples. It is validated for Western BlotSpecifications
| NPFF | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 8620 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| FMRFAL, FMRFamide-related peptides, neuropeptide FF-amide peptide precursor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 34-113 of human NPFF (NP_003708.1). AEEDSEPLPPQDAQTSGSLLHYLLQAMERPGRSQAFLFQPQRFGRNTQGSWRNEWLSPRAGEGLNSQFWSLAAPQRFGKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title