missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPHP3 Monoclonal Antibody (3B1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524451
This item is not returnable.
View return policy
Description
Peptide Sequence: NQELLSMGRR EAKLDTENKR LRAELQALQK TYQKILREKE SALEAKYQAM ERAATFEHDR DKVKRQFKIF RETKENEIQD LLRAKRELES KLQRLQAQGI.
Required for normal ciliary development and function. Inhibits disheveled-1-induced canonical Wnt-signaling activity and may also play a role in the control of non-canonical Wnt signaling which regulates planar cell polarity. Probably acts as a molecular switch between different Wnt signaling pathways. Required for proper convergent extension cell movements.
Specifications
| NPHP3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q7Z494 | |
| NPHP3 | |
| NPHP3 (NP_694972.3, 106 a.a. ~ 205 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3B1 | |
| Unconjugated | |
| NPHP3 | |
| 3632410F03Rik; AI550417; C230078J01; CFAP31; cilia and flagella associated protein 31; D330020E01Rik; Kiaa2000; Meckel syndrome, type 7; MKS7; Nephrocystin-3; nephronophthisis 3 (adolescent); NPH3; NPHP3; pcy; RHPD; RHPD1; SLSN3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 27031 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction