missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544883
This item is not returnable.
View return policy
Description
Peptide sequence: MNLSSASSTE EKAVTTVLWG CELSQERRTW TFRPQLEGKQ SCRLLLHTIC Sequence homology: Human: 100%.
NPM2 is a core histones chaperone involved in chromatin reprogramming, specially during fertilization and early embryonic development. It probably involved in sperm DNA decondensation during fertilization.
Specifications
| NPM2 | |
| Polyclonal | |
| Unconjugated | |
| NPM2 | |
| NPM2; nucleophosmin/nucleoplasmin 2; nucleoplasmin-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10361 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86SE8 | |
| NPM2 | |
| Synthetic peptide directed towards the N-terminal of human NPM2 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction