Learn More
NPPB Mouse anti-Human, Clone: OTI11C3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809079
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the natriuretic peptide family and encodes a secreted protein which functions as a cardiac hormone. The protein undergoes two cleavage events, one within the cell and a second after secretion into the blood. The protein′s biological actions include natriuresis, diuresis, vasorelaxation, inhibition of renin and aldosterone secretion, and a key role in cardiovascular homeostasis. A high concentration of this protein in the bloodstream is indicative of heart failure. Mutations in this gene have been associated with postmenopausal osteoporosis.Specifications
| NPPB | |
| Monoclonal | |
| Unconjugated | |
| NPPB | |
| BNP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4879 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI11C3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P16860 | |
| NPPB | |
| Human recombit protein fragment corresponding to amino acids 27-102 of human NPPB produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.