missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASGR2 Mouse anti-Human, Clone: OTI2A12, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505099S
This item is not returnable.
View return policy
Description
ASGR2 is a cell surface receptor that binds to galactose-terminated glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociates. Then the receptor is recycled back to the cell surface. There are four alternatively spliced transcript variants of this gene. This gene has multiple polyadenylation sites.Specifications
| ASGR2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07307 | |
| ASGR2 | |
| Full length human recombit protein of human ASGR2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A12 | |
| Unconjugated | |
| ASGR2 | |
| ASGP-R2, ASGPR2, CLEC4H2, HBXBP, HL-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 433 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction