missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPR3 Monoclonal Antibody (OTI4A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501046S
This item is not returnable.
View return policy
Description
NPR3 Monoclonal antibody specifically detects NPR3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| NPR3 | |
| Monoclonal | |
| 0.78 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17342 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4883 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG3 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4A1 | |
| Unconjugated | |
| NPR3 | |
| A NPR-C; ANP-C; ANP-CR; ANPRC; ANPR-C; atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide clearence receptor 3; atrial natriuretic peptide C-type receptor; atrial natriuretic peptide receptor 3; atrial natriuretic peptide receptor 3-like; atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; C5orf23; EF-2; guanylate cyclase C; GUCY2B; lgj; longjohn; natriuretic peptide receptor 3; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C); Nppc receptor; Npr3; NPRC; NPR-C; stri; strigosus | |
| Full length human recombinant protein of human NPR3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction