missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPR3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142824
This item is not returnable.
View return policy
Description
Additional Information: The immunizing peptide represents the cytoplasmic part of the protein. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
This gene encodes one of three natriuretic peptide receptors. Natriutetic peptides are small peptides which regulate blood volume and pressure, pulmonary hypertension, and cardiac function as well as some metabolic and growth processes. The product of this gene encodes a natriuretic peptide receptor responsible for clearing circulating and extracellular natriuretic peptides through endocytosis of the receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| NPR3 | |
| Polyclonal | |
| Unconjugated | |
| NPR3 | |
| A NPR-C; ANP-C; ANP-CR; ANPRC; ANPR-C; atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide clearence receptor 3; atrial natriuretic peptide C-type receptor; atrial natriuretic peptide receptor 3; atrial natriuretic peptide receptor 3-like; atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; C5orf23; EF-2; guanylate cyclase C; GUCY2B; lgj; longjohn; natriuretic peptide receptor 3; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C); Nppc receptor; Npr3; NPRC; NPR-C; stri; strigosus | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4883 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P17342 | |
| NPR3 | |
| Peptide with sequence C-HRELREDSIRSHFS. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction