missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPRB/NPR2 Antibody (1060722) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9725JF669
This item is not returnable.
View return policy
Description
NPRB/NPR2 Monoclonal antibody specifically detects NPRB/NPR2 in Human samples. It is validated for Flow Cytometry
Specifications
| NPRB/NPR2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| AMD1;AMDM;ANPb;ANPRB;Atrial natriuretic peptide receptor 2;ECDM;GCB;GC-B;GUC2B;GUCY2B;NPRB;NPRBi;SNSK | |
| Mouse myeloma cell line, NS0-derived human NPRB/NPR2, Arg23-Ile458, Accession # P20594 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 1060722 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction | |
| 4882 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction