missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPRL3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-97766
This item is not returnable.
View return policy
Description
NPRL3 Polyclonal specifically detects NPRL3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| NPRL3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| -14 gene protein, Alpha-globin regulatory element-containing gene protein, C16orf35, CGTHBAchromosome 16 open reading frame 35, conserved gene telomeric to alpha globin cluster, MAREHS-40, nitrogen permease regulator 3-like protein, nitrogen permease regu | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8131 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_853547.1 | |
| NPRL3 | |
| Peptide with sequence C-QRSQEHPASQTNKP corresponding to internal region (near N-Terminus) according to Uniprot Mouse NP_853547.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse and Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction