missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPRL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578245
This item is not returnable.
View return policy
Description
Positive Control: K562, U87-MG Predicted Reactivity: Mouse (97%), Rat (97%), Xenopus laevis (91%), Rabbit (97%), Bovine (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The function of the encoded protein is not known.
Specifications
| NPRL3 | |
| Polyclonal | |
| Unconjugated | |
| NPRL3 | |
| -14 gene protein; Aag; alpha-globin regulatory element-containing gene protein; alphaRE-14; C16orf35; CGTHBA; conserved gene telomeric to alpha globin cluster; GATOR complex protein NPRL3; HS-26; HS40; HS-40; MARE; nitrogen permease regulator 3-like protein; NPR3; NPR3 like, GATOR1 complex subunit; NPR3-like; NPR3-like, GATOR1 complex subunit; NPRL3; Phg; Protein CGTHBA; Prox1; RMD11; RP23-177M10.6 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8131 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.57 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q12980 | |
| NPRL3 | |
| Recombinant protein encompassing a sequence within the center region of human NPRL3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction