missing translation for 'onlineSavingsMsg'
Learn More

NPS-1034, MedChemExpress

Product Code. 16479616
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479616 50 mg 50mg
16485787 5 mg 5mg
16444356 100 mg 100mg
16442596 25 mg 25mg
16423058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16479616 Supplier Medchem Express Supplier No. HY100509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NPS-1034 is a dual inhibitor of AXL and MET with IC50s of 10.3 and 48 nM, respectively.

NPS-1034 is a dual inhibitor of AXL and MET with IC50s of 10.3 and 48 nM, respectively.

Specifications

Chemical Name or Material NPS-1034
CAS 1221713-92-3
Color White
Purity Grade Notes Research
Molecular Formula C31H23F2N5O3
Quantity 50 mg
Solubility Information DMSO : 34 mg/mL (61.65 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N(C)N(C2=CC=C(F)C=C2)C1=O)NC3=CC=C(OC4=C5C(NC=C5C6=CC=CC=C6)=NC=C4)C(F)=C3
Molecular Weight (g/mol) 551.54
Formula Weight 551.54
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.