missing translation for 'onlineSavingsMsg'
Learn More

NPS-2143, MedChemExpress

Product Code. 16455507
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16455507

Brand: Medchem Express HY10007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NPS-2143 (SB-262470A), an orally active calcilytic agent, is a selective and potent calcium ion-sensing receptor (CaSR) antagonist. NPS-2143 (SB-262470A) blocks increases in cytoplasmic Ca2+ concentrations (IC50=43 nM) elicited by activating the Ca2+ receptor in HEK 293 cells expressing the human Ca2+ receptor.

NPS-2143 (SB-262470A), an orally active calcilytic agent, is a selective and potent calcium ion-sensing receptor (CaSR) antagonist. NPS-2143 (SB-262470A) blocks increases in cytoplasmic Ca2+ concentrations (IC50=43 nM) elicited by activating the Ca2+ receptor in HEK 293 cells expressing the human Ca2+ receptor.

Specifications

Chemical Name or Material NPS-2143
CAS 284035-33-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25ClN2O2
Quantity 5 mg
Synonym SB-262470A
Solubility Information DMSO : 17.86 mg/mL (43.68 mM; ultrasonic and adjust pH to 2 with H2O)
SMILES N#CC1=C(OC[C@@H](CNC(C)(CC2=CC=C3C=CC=CC3=C2)C)O)C=CC=C1Cl
Molecular Weight (g/mol) 408.92
Formula Weight 408.92
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.