Learn More
NPTN Monoclonal Antibody (OTI2D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504330
Encompass_Preferred
 View more versions of this product
Product Code. 17581412
  • € 540.87 / 100µL
Description
Neuroplastin encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
NPTN
Monoclonal
0.69 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P97300, Q9Y639
NPTN
Full length human recombit protein of human NPTN produced in HEK293T cell.
100 μL
Primary
Canine, Human, Mouse
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2D4
Unconjugated
NPTN
GP55, GP65, SDFR1, SDR1, np55, np65
Mouse
Affinity Chromatography
RUO
20320, 27020, 487637
-20° C, Avoid Freeze/Thaw Cycles