missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPY1R Antibody (556153) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6400D
This item is not returnable.
View return policy
Description
NPY1R Monoclonal antibody specifically detects NPY1R in Human samples. It is validated for Flow Cytometry, CyTOF-reported
Specifications
| NPY1R | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 556153 | |
| Flow Cytometry, CyTOF-reported | |
| neuropeptide Y receptor type 1, neuropeptide Y receptor Y1, NPY1-R, NPYR, NPYY1 | |
| NS0 mouse myeloma cell line transfected with human NPY1R, Met1-Ile384, Accession # P25929 | |
| 0.1 mL | |
| GPCR, Neuroscience, Neurotransmission | |
| 4886 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction