missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPY1R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575330
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Neuropeptide Y receptors are a class of G-protein coupled receptors which are activated by the closely related peptide horomones neuropeptide Y, peptide YY and pancreatic polypeptide. These receptors are involved in control of a diverse set of behavioral processes including appetite, circadian rhythme, and anxiety.
Specifications
| NPY1R | |
| Polyclonal | |
| Unconjugated | |
| NPY1R | |
| FC5; Neuropepetide Y1 Receptor; neuropeptide Y receptor subtype; neuropeptide Y receptor type 1; neuropeptide Y receptor Y1; neuropeptide Y1 receptor; NPY-1; NPY-1 receptor; Npy1r; NPY1-R; NPYR; NPYY1; NPY-Y1 receptor; PY receptor Y1; Y1-R | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18166, 29358, 4886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P21555, P25929, Q04573 | |
| NPY1R | |
| Synthetic peptide corresponding to amino acids 225-270 of Human NPY1R. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction