missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPY2R Antibody (557521) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10211B
This item is not returnable.
View return policy
Description
NPY2R Monoclonal antibody specifically detects NPY2R in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| NPY2R | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 557521 | |
| Flow Cytometry, CyTOF-ready | |
| neuropeptide Y receptor type 2, neuropeptide Y receptor Y2, NPY2-R, NPY-Y2 receptor, Y2 receptor | |
| Mouse myeloma cell line NS0-derived transfected with recombinant human NPY2R, Met1-Val381, Accession # P49146 | |
| 0.1 mL | |
| GPCR, Neuroscience, Neurotransmission | |
| 4887 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction