missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPY5R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533991
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Marmoset, Bovine, Panda (94%) Monkey, Dog (88%).
Receptor for neuropeptide Y and peptide YY. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity. Seems to be associated with food intake. Could be involved in feeding disorders.
Specifications
| NPY5R | |
| Polyclonal | |
| Unconjugated | |
| Npy5r | |
| HGNC:7958; Neuropeptide Y receptor type 5; neuropeptide Y receptor type 5-like protein; neuropeptide Y receptor Y5; Neuropeptide Y5 receptor; Npy5; NPY5 receptor; Npy5r; NPY5-R; NPYR5; NPYY5; NPY-Y5 receptor; NPYY5-R; Y5 receptor; Y5R | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4889 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q15761 | |
| Npy5r | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human NPY5R. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction