missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQDI-1, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | NQDI-1 |
| CAS | 175026-96-7 |
| Molecular Formula | C19H13NO4 |
| Quantity | 5 mg |
| Synonym | Ethyl 2,7-Dioxo-3,7-dihydro-2H-naphtho[1,2,3-de]quinoline-1-carboxylate, 2-Hydroxy-7-oxo-7H-naphtho[1,2,3-de]quinoline-1-carboxylic Acid Ethyl Ester, 2,7-Dihydro-2,7-dioxo-3H-naphtho[1,2,3-de]quinoline-1-carboxylic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C19H13NO4/c1-2-24-19(23)16-15-10-6-3-4-7-11(10)17(21)12-8-5-9-13(14(12)15)20-18(16)22/h3-9H,2H2,1H3,(H,20,22) |
| SMILES | O=C(C(C(NC1=CC=CC2=C31)=O)=C3C4=C(C=CC=C4)C2=O)OCC |
| IUPAC Name | ethyl 2,7-dioxo-2,7-dihydro-3H-naphtho[1,2,3-de]quinoline-1-carboxylate |
| Molecular Weight (g/mol) | 319.31 |
| Formula Weight | 319.08 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction