missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO-2 Antibody (OTI3G4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73049APC
This item is not returnable.
View return policy
Description
NQO-2 Monoclonal antibody specifically detects NQO-2 in Human, Mouse, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| NQO-2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Canine, Primate, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI3G4 | |
| Western Blot, Immunohistochemistry | |
| dioxin-inducible-2, EC 1.10.99.2, NAD(P)H dehydrogenase, quinone 2, NAD(P)H menadione oxidoreductase 2, dioxin-inducible, NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, ribosyldihydronicotinamide dehydrogenase, ribosyldihydronicotinamide dehydrogenase [quinone] | |
| Full length human recombinant protein of human NQO2(NP_000895) produced in HEK293T cell. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 4835 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction