Learn More
NR0B2 Mouse anti-Human, Clone: OTI6D8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806318
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is an unusual orphan receptor that contains a putative ligand-binding domain but lacks a conventional DNA-binding domain. The gene product is a member of the nuclear hormone receptor family, a group of transcription factors regulated by small hydrophobic hormones, a subset of which do not have known ligands and are referred to as orphan nuclear receptors. The protein has been shown to interact with retinoid and thyroid hormone receptors, inhibiting their ligand-dependent transcriptional activation. In addition, interaction with estrogen receptors has been demonstrated, leading to inhibition of function. Studies suggest that the protein represses nuclear hormone receptor-mediated transactivation via two separate steps: competition with coactivators and the direct effects of its transcriptional repressor function.Specifications
| NR0B2 | |
| Monoclonal | |
| Unconjugated | |
| NR0B2 | |
| SHP, SHP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8431 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6D8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q15466 | |
| NR0B2 | |
| Full length human recombit protein of human NR0B2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.