missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NR1D1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15352711
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15352711 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15352711 Supplier Invitrogen™ Supplier No. PA533997

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Elephant, Panda, Dog, Rabbit (100%) Gorilla, Gibbon, Marmoset, Bat, Horse, Pig (93%) Monkey, Mouse, Bovine (87%).

This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR1D1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NR1D1
Gene Accession No. P20393
Gene Alias A530070C09Rik; BD73; Ear1; ear-1; EAR-1R; HREV; NR1D1; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; R75201; rev-erb alpha; Rev-erba; rev-erbA alpha; rev-erbA(alpha); Reverbaalpha; Rev-ErbAalpha; rev-erbA-alpha; Rev-ErbA-alpha protein; REV-ERBalpha; RVR; THRA1; THRAL; V-erbA-related protein 1; V-erbA-related protein EAR-1
Gene Symbols NR1D1
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from internal region of human NR1D1.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100357678, 100688236, 9572
Target Species Human, Canine, Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.