missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1D1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14506-1-AP-150UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against the N terminus of human NR1D1.
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.Specifications
| NR1D1 | |
| Polyclonal | |
| Unconjugated | |
| NR1D1 | |
| ear 1, EAR1, hRev, NR1D1, Rev erbA alpha, Reverb alpha, THRA1, THRAL, V erbA related protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217166, 9572 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P20393, Q3UV55 | |
| NR1D1 | |
| NR1D1 Fusion Protein Ag5964 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction