missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NR1D2 Polyclonal Antibody

Product Code. 13444619
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13444619 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13444619 Supplier Invitrogen™ Supplier No. PA532948

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Bat, Panda, Horse, Rabbit, Turkey, Chicken (100%); Elephant, Platypus (94%); Opossum, Xenopus (81%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR1D2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NR1D2
Gene Accession No. Q14995, Q60674, Q63504
Gene Alias BD73; EAR-1R; EAR4; HZF2; HZF-2; Nr1d2; nuclear receptor Rev-ErbA beta variant 1; nuclear receptor Rev-ErbA beta variant 2; nuclear receptor subfamily 1 group D member 2; nuclear receptor subfamily 1, group D, member 2; orphan nuclear hormone receptor BD73; orphan nuclear receptor HZF-2; orphan nuclear receptor RVR; orphan receptor; Rev-erb; Rev-erb alpha-related receptor; Rev-erb beta; Rev-erba beta; rev-erba-alpha-related receptor; Rev-erbabeta; Rev-ErbA-beta; rev-erb-beta; Rev-erb-Beta2; RVR; V-erbA-related protein 1-related
Gene Symbols NR1D2
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human NR1D2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100060122, 100349421, 259241, 353187, 396125, 485643, 532076, 9975
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Horse, Chicken, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.