missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1I3 Monoclonal Antibody (OTI8E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805367S
This item is not returnable.
View return policy
Description
NR1I3 Monoclonal antibody specifically detects NR1I3 in Human samples. It is validated for Western BlotSpecifications
| NR1I3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14994 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9970 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI8E3 | |
| Unconjugated | |
| NR1I3 | |
| AA209988; AI551208; Car; CAR1; CAR-beta; Care2; constitutive activator of retinoid response; constitutive active receptor; constitutive active response; constitutive androstane nuclear receptor variant 2; constitutive androstane nuclear receptor variant 3; constitutive androstane nuclear receptor variant 4; constitutive androstane nuclear receptor variant 5; constitutive androstane receptor; constitutive androstane receptor NR1I3; constitutive androstane receptor protein variant 1; constitutive androstane receptor protein variant 2; constitutive androstane receptor protein variant 3; constitutive androstane receptor protein variant 4; constitutive androstane receptor protein variant 5; constitutively transactivates RAREs; ESTM32; MB67; MGC150433; MGC97144; MGC97209; NR1I3; nuclear receptor; nuclear receptor constitutive active receptor; nuclear receptor subfamily 1 group I member 3; nuclear receptor subfamily 1, group I, member 3; orphan nuclear hormone receptor; orphan nuclear recepto | |
| Human recombinant protein fragment corresponding to amino acids 1-236 of human NR1I3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction