missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2C1 Mouse anti-Human, Clone: OTI4C7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803381
This item is not returnable.
View return policy
Description
This gene encodes a nuclear hormone receptor characterized by a highly conserved DNA binding domain (DBD), a variable hinge region, and a carboxy-terminal ligand binding domain (LBD) that is typical for all members of the steroid/thyroid hormone receptor superfamily. This protein also belongs to a large family of ligand-inducible transcription factors that regulate gene expression by binding to specific DNA sequences within promoters of target genes. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these variants has not been determined.Specifications
| NR2C1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P13056 | |
| NR2C1 | |
| Human recombit protein fragment corresponding to amino acids 1-237 of human NR2C1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI4C7 | |
| Unconjugated | |
| NR2C1 | |
| TR2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7181 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction