missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2C2 Monoclonal Antibody (OTI1D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807248S
This item is not returnable.
View return policy
Description
NR2C2 Monoclonal antibody specifically detects NR2C2 in Human samples. It is validated for Western BlotSpecifications
| NR2C2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49116 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7182 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1D6 | |
| Unconjugated | |
| NR2C2 | |
| mKIAA4145; Mtr2r1; Nr2c2; Nuclear hormone receptor TR4; nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; nuclear receptor subfamily 2, group H, member 2; orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; orphan receptor, TR4; Tak1; testicular nuclear receptor 4; testicular receptor 4; TR2R1; Tr4; TR4 orphan receptor; TR4-NS orphan receptor | |
| Human recombinant protein fragment corresponding to amino acids 1-241 of human NR2C2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction