missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2C2 Monoclonal Antibody (OTI7E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807249S
This item is not returnable.
View return policy
Description
NR2C2 Monoclonal antibody specifically detects NR2C2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NR2C2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49116 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7182 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7E12 | |
| Unconjugated | |
| NR2C2 | |
| mKIAA4145; Mtr2r1; Nr2c2; Nuclear hormone receptor TR4; nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; nuclear receptor subfamily 2, group H, member 2; orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; orphan receptor, TR4; Tak1; testicular nuclear receptor 4; testicular receptor 4; TR2R1; Tr4; TR4 orphan receptor; TR4-NS orphan receptor | |
| Human recombinant protein fragment corresponding to amino acids 1-241 of human NR2C2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion