missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2E3 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806219S
This item is not returnable.
View return policy
Description
NR2E3 Monoclonal antibody specifically detects NR2E3 in Human samples. It is validated for Western BlotSpecifications
| NR2E3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5X4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10002 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G3 | |
| Unconjugated | |
| NR2E3 | |
| A930035N01Rik; ESCS; MGC138414; MGC138416; Nr2e3; Nuclear receptor subfamily 2 group E member 3; nuclear receptor subfamily 2, group E, member 3; photoreceptor-specific nuclear receptor; PNR; rd7; retinal degeneration 7; Retina-specific nuclear receptor; Rnr; RP11-295F4.5; RP37; taR-5 | |
| Full length human recombinant protein of human NR2E3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction