missing translation for 'onlineSavingsMsg'
Learn More

NR2F6 Monoclonal Antibody (OTI2C9), TrueMAB™, OriGene

Product Code. p-200013682
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17596993 30 μL 30µL
17576993 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17596993 Supplier Origene Technologies Supplier No. TA806769S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NR2F6 Monoclonal antibody specifically detects NR2F6 in Human samples. It is validated for Western Blot

Specifications

Antigen NR2F6
Applications Western Blot
Classification Monoclonal
Clone OTI2C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FLT1
Gene Accession No. P17948
Gene Alias AV090102; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 2; COUP transcription factor 3; COUPg; COUP-TF3; EAR2; EAR-2; Erba related 2; ERBAL2; ERBA-related gene-2; Nr2f6; Nuclear receptor subfamily 2 group F member 6; nuclear receptor subfamily 2, group F, member 6; nuclear receptor V-erbA-related; orphan nuclear receptor EAR-2; Ovalbumin upstream promoter gamma nuclear receptor; ovalbumin upstream promoter gamma nuclear receptor rCOUPg; v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2; V-erbA-related protein 2; V-erbA-related protein EAR-2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 781-1338 of human FLT1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2321
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.