missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR4A3 Mouse anti-Human, Clone: OTI2A6, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804855S
This item is not returnable.
View return policy
Description
NR4A3 Monoclonal antibody specifically detects NR4A3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NR4A3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92570 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8013 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A6 | |
| Unconjugated | |
| NR4A3 | |
| AI573420; CHN; chondrosarcoma, extraskeletal myxoid, fused to EWS; CSMF; MINOR; mitogen-induced nuclear orphan receptor; Neuron-derived orphan receptor 1; neuron-derived orphan receptor 1/2; Nor1; NOR-1; NOR-2; Nr4a3; Nuclear hormone receptor NOR-1; nuclear hormone receptor NOR-1/NOR-2; Nuclear receptor subfamily 4 group A member 3; nuclear receptor subfamily 4, group A, member 3; Orphan nuclear receptor TEC; TEC; TEC receptor; Translocated in extraskeletal chondrosarcoma | |
| Human recombinant protein fragment corresponding to amino acids 332-626 of human NR4A3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction