missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR4A3/NOR1 Antibody (OTI5C2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73052AF405
This item is not returnable.
View return policy
Description
NR4A3/NOR1 Monoclonal specifically detects NR4A3/NOR1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NR4A3/NOR1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NR4A3 | |
| Recombinant protein fragment corresponding to amino acids 332-626 of human NR4A3/NOR1 (NP_008912) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI5C2 | |
| Western Blot | |
| CHN, CSMF, MINOR, NOR1, TEC | |
| Mouse | |
| Affinity Purified | |
| GPCR | |
| 8013.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction