missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR5A2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143112
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_995582.1; NP_003813.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Binds to the sequence element 5'-AACGACCGACCTTGAG-3' of the enhancer II of hepatitis B virus genes, a critical cis-element of their expression and regulation. May be responsible for the liver-specific activity of enhancer II, probably in combination with other hepatocyte transcription factors. Key regulator of cholesterol 7-alpha-hydroxylase gene (CYP7A) expression in liver. May also contribute to the regulation of pancreas-specific genes and play important roles in embryonic development.
Specifications
| NR5A2 | |
| Polyclonal | |
| Unconjugated | |
| Nr5a2 | |
| Alpha-1-fetoprotein transcription factor; AU020803; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; B1F; B1F2; CPF; CYP7A promoter-binding factor; D1Ertd308e; fetoprotein transcription factor; fetoprotein-alpha 1 (AFP) transcription factor; Ftf; FTF/LRH-1; FTZ F1; FTZ F1beta; FTZ-F1; FTZ-F1 beta; FTZ-F1 beta2 protein; FTZ-F1beta; hB1F; hB1F 2; hB1F-2; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; I79_021747; live; liver nuclear receptor homolog-1 variant 2; liver receptor homolog 1; liver receptor homolog-1; Lrh1; LRH-1; Nr5a2; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2; OR2.0; orphan nuclear receptor; SF-2; steroidogenic factor 2; UF2-H3B | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 100523688, 2494 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O00482 | |
| Nr5a2 | |
| Peptide with sequence C-TDYDRSPFVTSPIS. | |
| 100 μg | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction