missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NR5A2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110041

Product Code. 17137973

  • € 493.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SIPHLILELL KCEPDEPQVQ AKIMAYLQQE QANRSKHEKL STFGLMCKMA DQTLFSIVEW ARSSIFFREL KVDDQMKLLQ NCWSELLILD HIYRQVVHGK EGSIFLVTGQ QVDYSIIASQ AGATLNNLMS HAQELVAKLR SLQFDQREFV CLKFLVLFSL DVKNLENFQL VEGVQEQVNA ALLDYTMCNY PQQTEKFGQL LLRLPEIRAI SMQAEEYLYY KHLNGDVPYN NLLIEMLHAK RA.

Binds to the sequence element 5'-AACGACCGACCTTGAG-3' of the enhancer II of hepatitis B virus genes, a critical cis-element of their expression and regulation. May be responsible for the liver-specific activity of enhancer II, probably in combination with other hepatocyte transcription factors. Key regulator of cholesterol 7-alpha-hydroxylase gene (CYP7A) expression in liver. May also contribute to the regulation of pancreas-specific genes and play important roles in embryonic development.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NR5A2
Polyclonal
Unconjugated
Nr5a2
Alpha-1-fetoprotein transcription factor; AU020803; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; B1F; B1F2; CPF; CYP7A promoter-binding factor; D1Ertd308e; fetoprotein transcription factor; fetoprotein-alpha 1 (AFP) transcription factor; Ftf; FTF/LRH-1; FTZ F1; FTZ F1beta; FTZ-F1; FTZ-F1 beta; FTZ-F1 beta2 protein; FTZ-F1beta; hB1F; hB1F 2; hB1F-2; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; I79_021747; live; liver nuclear receptor homolog-1 variant 2; liver receptor homolog 1; liver receptor homolog-1; Lrh1; LRH-1; Nr5a2; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2; OR2.0; orphan nuclear receptor; SF-2; steroidogenic factor 2; UF2-H3B
Rabbit
Affinity chromatography
RUO
2494, 26424, 60349
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.81 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00482, P45448, Q9QWM1
Nr5a2
Recombinant fusion protein containing a sequence corresponding to amino acids 300-541 of human NR5A2 (NP_9955821).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.