missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRAGE/MAGED1 Antibody (353124), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3835MFV610
This item is not returnable.
View return policy
Description
NRAGE/MAGED1 Monoclonal antibody specifically detects NRAGE/MAGED1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| NRAGE/MAGED1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 9500 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 353124 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| DLXIN-1, MAGE tumor antigen CCF, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen | |
| E. coli-derived recombinant human NRAGE, Met1-Glu778, Accession # Q9Y5V3 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction