missing translation for 'onlineSavingsMsg'
Learn More

NRAS Antibody (06), Novus Biologicals™

Mouse Monoclonal Antibody

€ 231.00 - € 326.00

Specifications

Antigen NRAS
Clone 06
Dilution Western Blot 1:500-1:2000, ELISA 1:1000-1:2000
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18699187
View Documents
Novus Biologicals
NBP3-05798-100ul
100 μg
€ 326.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18611229
View Documents
Novus Biologicals
NBP3-05798-50ul
50 μg
€ 231.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NRAS Monoclonal antibody specifically detects NRAS in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

NRAS
Western Blot 1:500-1:2000, ELISA 1:1000-1:2000
Monoclonal
Purified
RUO
Human
ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NRAS (Uniprot#: P01111; Met1-Cys186).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
06
Western Blot, ELISA
Unconjugated
Mouse
Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway
0.2 um filtered solution in PBS
4893
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.