missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRAS Antibody (OTI5G7), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73053V
This item is not returnable.
View return policy
Description
NRAS Monoclonal specifically detects NRAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| NRAS | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NRAS | |
| Full length recombinant protein of human NRAS (NP_002515) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5G7 | |
| Western Blot, Immunohistochemistry | |
| ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog | |
| Mouse | |
| Affinity Purified | |
| Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway | |
| 4893.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction