missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NRAS Monoclonal antibody specifically detects NRAS in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | NRAS |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S01-7D0 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human NRAS (Uniprot # P01111) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?